siRNA Details
Gene Symbol: MKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308142.1 | NP_001295071.1 | 6 | 504 | ||
NM_014048.4 | NP_054767.3 | 6 | 505 | |||
XM_005255452.3 | XP_005255509.1 | 7 | 659 | |||
XM_005255453.4 | XP_005255510.1 | 6 | 1143 | |||
XM_005255455.3 | XP_005255512.1 | 4 | 489 | |||
XM_006720908.3 | XP_006720971.1 | 7 | 1305 | |||
XM_006720909.3 | XP_006720972.1 | 7 | 634 | |||
XM_006720912.3 | XP_006720975.1 | 4 | 2444 | |||
XM_006720913.2 | XP_006720976.1 | 7 | 659 | |||
XM_006720914.2 | XP_006720977.1 | 7 | 659 | |||
XM_011522568.2 | XP_011520870.1 | 6 | 474 | |||
XM_011522569.2 | XP_011520871.1 | 4 | 271 | |||
XM_017023501.1 | XP_016878990.1 | 6 | 470 | |||
XM_017023502.1 | XP_016878991.1 | 6 | 527 | |||
XM_017023503.1 | XP_016878992.1 | 5 | 3672 | |||
XM_017023504.1 | XP_016878993.1 | 4 | 489 | |||
XM_017023505.1 | XP_016878994.1 | 4 | 2444 | |||
GenBank mRNA | AK093577.1 | - | 4 | 489 | ||
AK294765.1 | - | 6 | 496 | |||
AY374297.1 | - | 6 | 482 | |||
BC034934.1 | - | 5 | 457 | |||
BC047761.1 | - | 4 | 329 | |||
BC127941.1 | - | 4 | 210 | |||
BC136260.1 | - | 4 | 349 | 9020247 | ||
BC171750.1 | - | 4 | 349 | 9053251 | ||
DB114294.1 | - | 6 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
