siRNA Details
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001424.5 | NP_001415.1 | 5 | 910 | ||
XM_006720864.3 | XP_006720927.1 | 5 | 904 | |||
GenBank mRNA | AK096403.1 | - | 1 | 34 | ||
BM755118.1 | - | 1 | 32 | |||
BQ676037.1 | - | 152 | ||||
BU838264.1 | - | 1 | 152 | |||
DA752772.1 | - | 1 | 34 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
