siRNA Details
Gene Symbol: CCDC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080850.2 | NP_001074319.1 | 6, 7 | 972 | ||
XM_011542067.2 | XP_011540369.1 | 7, 8 | 1904 | |||
XM_011542068.2 | XP_011540370.1 | 7, 8 | 1909 | |||
XM_011542069.2 | XP_011540371.1 | 6, 7 | 1763 | |||
XM_011542070.2 | XP_011540372.1 | 5, 6 | 930 | |||
XM_011542071.2 | XP_011540373.1 | 4, 5 | 1339 | |||
XM_017002225.1 | XP_016857714.1 | 6, 7 | 989 | |||
XM_017002226.1 | XP_016857715.1 | 7, 8 | 1300 | |||
GenBank mRNA | AK302075.1 | - | 8, 9 | 1156 | ||
AL832217.1 | - | 13, 14 | 2005 | |||
AY639646.1 | - | 5, 6 | 797 | |||
BX640642.1 | - | 2, 3 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
