siRNA Details
Gene Symbol: CCDC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080850.2 | NP_001074319.1 | 7 | 1000 | ||
XM_011542067.2 | XP_011540369.1 | 8 | 1932 | |||
XM_011542068.2 | XP_011540370.1 | 8 | 1937 | |||
XM_011542069.2 | XP_011540371.1 | 7 | 1791 | |||
XM_011542070.2 | XP_011540372.1 | 6 | 958 | |||
XM_011542071.2 | XP_011540373.1 | 5 | 1367 | |||
XM_017002225.1 | XP_016857714.1 | 7 | 1017 | |||
XM_017002226.1 | XP_016857715.1 | 8 | 1328 | |||
GenBank mRNA | AK302075.1 | - | 9 | 1184 | ||
AL832217.1 | - | 14 | 2033 | |||
AY639646.1 | - | 6 | 825 | |||
BX640642.1 | - | 3 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
