siRNA Details
Gene Symbol: ZFP91-CNTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024091.1 | 7 | 1058 | |||
GenBank mRNA | AB057443.1 | - | 7 | 1004 |
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197051.1 | NP_001183980.1 | 7 | 1055 | ||
NM_053023.4 | NP_444251.1 | 7 | 1058 | |||
GenBank mRNA | AB056107.1 | - | 7 | 1004 | ||
AF310246.1 | - | 7 | 704 | |||
BC051743.1 | - | 7 | 1058 | 4827133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
