siRNA Details
Gene Symbol: ZNF721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133474.3 | NP_597731.2 | 3 | 574 | ||
GenBank mRNA | AK127067.1 | - | 1 | 130 | ||
AK128499.1 | - | 1 | 403 | |||
AL834155.1 | - | 378 | ||||
DA309597.1 | - | 3 | 557 |
Gene Symbol: ZNF732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 550 | ||
GenBank mRNA | AK302099.1 | - | 3 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
