siRNA Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018660.2 | NP_061130.1 | 10 | 1938 | ||
GenBank mRNA | AB044752.1 | - | 2 | 472 | ||
AK098243.1 | - | 16 | 2821 | |||
BG249131.1 | - | 2 | 492 | |||
BG759384.1 | - | 4 | 662 | |||
BI917232.1 | - | 3 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
