siRNA Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018660.2 | NP_061130.1 | 10 | 1994 | ||
GenBank mRNA | AB044752.1 | - | 2 | 528 | ||
AK023418.1 | - | 10 | 1994 | |||
AK098243.1 | - | 16 | 2877 | |||
BG249131.1 | - | 2 | 548 | |||
BI917232.1 | - | 3 | 612 | |||
CB960515.1 | - | 1 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
