siRNA Details
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204300.1 | NP_001191229.1 | 5 | 833 | ||
NM_001256423.1 | NP_001243352.1 | 5 | 816 | |||
NM_032900.5 | NP_116289.4 | 5 | 833 | |||
GenBank mRNA | AA427792.1 | - | 2 | 254 | ||
AK090447.1 | - | 5 | 783 | |||
AK093316.1 | - | 3 | 464 | |||
AK093441.1 | - | 5 | 790 | |||
AK303055.1 | - | 5 | 794 | |||
BC113888.1 | - | 5 | 809 | 40082328 | ||
BC114490.1 | - | 5 | 809 | 40082327 | ||
BX648189.1 | - | 5 | 808 | |||
BX648194.1 | - | 2 | 276 | |||
CR749412.1 | - | 5 | 809 | |||
DQ338460.1 | - | 5 | 798 |
Gene Symbol: ARHGAP19-SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037909.1 | 5 | 833 | |||
GenBank mRNA | AY336750.1 | - | 5 | 798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
