siRNA Details
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204300.1 | NP_001191229.1 | 11 | 2322 | ||
NM_001256423.1 | NP_001243352.1 | 12 | 2392 | |||
NM_032900.5 | NP_116289.4 | 12 | 2409 | |||
GenBank mRNA | AK074122.1 | - | 7 | 1489 | ||
AK090447.1 | - | 13 | 2454 | |||
BC113888.1 | - | 11 | 2298 | 40082328 | ||
BC114490.1 | - | 12 | 2386 | 40082327 | ||
BX648189.1 | - | 12 | 2384 | |||
BX648194.1 | - | 9 | 1851 | |||
CR749412.1 | - | 12 | 2385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
