siRNA Details
Gene Symbol: MCM3AP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002776.4 | 2 | 159 | |||
NR_110565.1 | 3 | 360 | ||||
NR_110566.1 | 3 | 262 | ||||
NR_110567.1 | 2 | 253 | ||||
GenBank mRNA | BC016306.2 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
