siRNA Details
Gene Symbol: HSPA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278205.1 | NP_001265134.1 | 2 | 360 | ||
NM_016299.3 | NP_057383.2 | 2 | 360 | |||
NR_103464.1 | 2 | 360 | ||||
GenBank mRNA | AF112210.1 | - | 2 | 179 | ||
AF143723.1 | - | 2 | 169 | |||
AK000880.1 | - | 2 | 201 | |||
AK225265.1 | - | 2 | 204 | |||
AK225651.1 | - | 2 | 204 | |||
AK292323.1 | - | 2 | 168 | |||
AK302452.1 | - | 2 | 204 | |||
BC065281.1 | - | 2 | 191 | |||
BC108738.1 | - | 2 | 294 | |||
BC119690.1 | - | 2 | 179 | 40116697 | ||
BP201211.1 | - | 2 | 360 | |||
BQ424016.1 | - | 2 | 291 | |||
CN308284.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
