siRNA Details
Gene Symbol: HSPA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278205.1 | NP_001265134.1 | 1 | 253 | ||
NM_016299.3 | NP_057383.2 | 1 | 253 | |||
NR_103464.1 | 1 | 253 | ||||
GenBank mRNA | AF112210.1 | - | 1 | 72 | ||
AF143723.1 | - | 1 | 62 | |||
AK000880.1 | - | 1 | 94 | |||
AK225265.1 | - | 1 | 97 | |||
AK225651.1 | - | 1 | 97 | |||
AK302452.1 | - | 1 | 97 | |||
BC065281.1 | - | 1 | 84 | |||
BC108738.1 | - | 1 | 187 | |||
BC119690.1 | - | 1 | 72 | 40116697 | ||
BP201211.1 | - | 1 | 253 | |||
BQ424016.1 | - | 1 | 184 | |||
CN308284.1 | - | 1 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
