siRNA Details
Gene Symbol: IFT140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014714.3 | NP_055529.2 | 16 | 2209 | ||
XM_005255725.4 | XP_005255782.1 | 15 | 1911 | |||
XM_005255726.3 | XP_005255783.1 | 15 | 1911 | |||
XM_006720990.3 | XP_006721053.1 | 15 | 2127 | |||
XM_006720991.3 | XP_006721054.1 | 15 | 2317 | |||
XM_011522766.2 | XP_011521068.1 | 13 | 1665 | |||
XM_011522767.2 | XP_011521069.1 | 8 | 929 | |||
XM_011522769.2 | XP_011521071.1 | 15 | 1911 | |||
XM_011522771.2 | XP_011521073.1 | 15 | 1911 | |||
XM_011522772.2 | XP_011521074.1 | 15 | 1911 | |||
XM_017023910.1 | XP_016879399.1 | 15 | 1900 | |||
XM_017023911.1 | XP_016879400.1 | 13 | 1760 | |||
GenBank mRNA | AB011162.1 | - | 15 | 1906 | ||
AK299035.1 | - | 15 | 1908 | |||
AK299115.1 | - | 10 | 1083 | |||
BC035577.1 | - | 14 | 1894 | 5552740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
