siRNA Details
Gene Symbol: TET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287491.1 | NP_001274420.1 | 11 | 4317 | ||
XM_005264187.3 | XP_005264244.1 | 12 | 4837 | |||
XM_011532682.2 | XP_011530984.1 | 13 | 4885 | |||
XM_011532683.2 | XP_011530985.1 | 13 | 5194 | |||
XM_011532684.2 | XP_011530986.1 | 13 | 6626 | |||
XM_011532685.2 | XP_011530987.1 | 12 | 4828 | |||
XM_011532686.2 | XP_011530988.1 | 11 | 4780 | |||
XM_011532687.2 | XP_011530989.1 | 12 | 4167 | |||
XM_011532689.2 | XP_011530991.1 | 11 | 1946 | |||
XM_011532690.1 | XP_011530992.1 | 10 | 2003 | |||
XM_017003566.1 | XP_016859055.1 | 13 | 6725 | |||
XM_017003567.1 | XP_016859056.1 | 11 | 4149 | |||
GenBank mRNA | AB007861.1 | - | 1 | 314 | ||
BC022243.1 | - | 8 | 1773 | 4400878 | ||
HQ220209.1 | - | 11 | 4317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
