siRNA Details
Gene Symbol: EXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017820.4 | NP_060290.3 | 9 | 974 | ||
XM_011518810.1 | XP_011517112.1 | 11 | 1304 | |||
XM_011518814.1 | XP_011517116.1 | 3 | 163 | |||
XM_017014861.1 | XP_016870350.1 | 12 | 1448 | |||
XM_017014862.1 | XP_016870351.1 | 10 | 1189 | |||
XM_017014863.1 | XP_016870352.1 | 10 | 1189 | |||
XM_017014864.1 | XP_016870353.1 | 9 | 1509 | |||
XM_017014865.1 | XP_016870354.1 | 9 | 1156 | |||
XM_017014866.1 | XP_016870355.1 | 8 | 812 | |||
GenBank mRNA | AK094438.1 | - | 9 | 874 | ||
AL536907.3 | - | 9 | 844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
