siRNA Details
Gene Symbol: TP73-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033708.1 | 1 | 406 | |||
NR_033709.1 | 1 | 406 | ||||
NR_033710.1 | 1 | 406 | ||||
NR_033711.1 | 1 | 406 | ||||
GenBank mRNA | AB007964.1 | - | 1 | 356 | ||
BC035073.1 | - | 1 | 371 | |||
BC089039.1 | - | 1 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
