siRNA Details
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001524.2 | NP_001001524.2 | 2 | 214 | ||
GenBank mRNA | AF255923.1 | - | 2 | 163 | ||
AK026307.1 | - | 2 | 205 | |||
AK097070.1 | - | 2 | 214 | |||
BC120986.1 | - | 2 | 188 | |||
BC120987.1 | - | 2 | 188 | |||
DA930032.1 | - | 2 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
