siRNA Details
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001524.2 | NP_001001524.2 | 3 | 279 | ||
GenBank mRNA | AF255923.1 | - | 3 | 228 | ||
AK097070.1 | - | 3 | 279 | |||
BC120986.1 | - | 3 | 253 | |||
BC120987.1 | - | 3 | 253 | |||
DA930032.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
