siRNA Details
Gene Symbol: SPRY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127496.1 | NP_001120968.1 | 2 | 720 | ||
NM_001293289.1 | NP_001280218.1 | 3 | 1098 | |||
NM_001293290.1 | NP_001280219.1 | 2 | 1036 | |||
NM_030964.3 | NP_112226.2 | 3 | 771 | |||
XM_011537685.2 | XP_011535987.1 | 3 | 1498 | |||
XM_017009910.1 | XP_016865399.1 | 2 | 1764 | |||
GenBank mRNA | AF227516.1 | - | 3 | 717 | ||
AK096464.1 | - | 2 | 705 | |||
AK226147.1 | - | 1 | 1143 | |||
BC125095.1 | - | 2 | 569 | 40118267 | ||
BC125096.1 | - | 2 | 569 | 40118268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
