siRNA Details
Gene Symbol: LRRC37A11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033753.2 | 1 | 485 | |||
GenBank mRNA | AK125814.1 | - | 1 | 484 | ||
BC111725.1 | - | 1 | 213 | 40028479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
