siRNA Details
Gene Symbol: DNAH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173628.3 | NP_775899.3 | 55 | 8662 | ||
XM_011525416.2 | XP_011523718.1 | 55 | 8698 | |||
XM_017025261.1 | XP_016880750.1 | 13 | 1922 | |||
GenBank mRNA | AK128065.1 | - | 2 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
