siRNA Details
Gene Symbol: FANCM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308133.1 | NP_001295062.1 | 7 | 1395 | ||
NM_001308134.1 | NP_001295063.1 | 8 | 1473 | |||
NM_020937.3 | NP_065988.1 | 8 | 1473 | |||
XM_011537034.2 | XP_011535336.1 | 8 | 1472 | |||
XM_011537035.2 | XP_011535337.1 | 7 | 1394 | |||
XM_017021523.1 | XP_016877012.1 | 8 | 1472 | |||
XM_017021524.1 | XP_016877013.1 | 6 | 694 | |||
XM_017021525.1 | XP_016877014.1 | 6 | 1261 | |||
XM_017021526.1 | XP_016877015.1 | 5 | 505 | |||
XM_017021527.1 | XP_016877016.1 | 7 | 1286 | |||
GenBank mRNA | AK093422.1 | - | 8 | 1472 | ||
AL833656.1 | - | 2 | 135 | |||
BC036056.1 | - | 8 | 1474 | |||
BC140776.1 | - | 7 | 1380 | 9021644 | ||
BC144511.1 | - | 7 | 1380 | 9053038 | ||
DQ140356.1 | - | 8 | 1480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
