siRNA Details
Gene Symbol: PEAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080471.1 | NP_001073940.1 | 9 | 1124 | ||
XM_005245141.3 | XP_005245198.1 | 10 | 1317 | |||
XM_011509510.2 | XP_011507812.1 | 10 | 1256 | |||
XM_011509511.2 | XP_011507813.1 | 9 | 1212 | |||
XM_011509512.2 | XP_011507814.1 | 9 | 1136 | |||
XM_017001234.1 | XP_016856723.1 | 9 | 2382 | |||
XM_017001235.1 | XP_016856724.1 | 9 | 2252 | |||
XM_017001236.1 | XP_016856725.1 | 9 | 1219 | |||
XM_017001237.1 | XP_016856726.1 | 10 | 1463 | |||
XM_017001238.1 | XP_016856727.1 | 10 | 1402 | |||
XM_017001239.1 | XP_016856728.1 | 10 | 1213 | |||
XM_017001240.1 | XP_016856729.1 | 7 | 781 | |||
XM_017001241.1 | XP_016856730.1 | 7 | 929 | |||
XM_017001242.1 | XP_016856731.1 | 7 | 833 | |||
GenBank mRNA | AK074121.1 | - | 4 | 737 | ||
AK098809.1 | - | 5 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
