siRNA Details
Gene Symbol: PEAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080471.1 | NP_001073940.1 | 7 | 816 | ||
XM_005245141.3 | XP_005245198.1 | 8 | 1009 | |||
XM_011509510.2 | XP_011507812.1 | 8 | 948 | |||
XM_011509511.2 | XP_011507813.1 | 7 | 904 | |||
XM_011509512.2 | XP_011507814.1 | 7 | 828 | |||
XM_017001234.1 | XP_016856723.1 | 7 | 2074 | |||
XM_017001235.1 | XP_016856724.1 | 7 | 1944 | |||
XM_017001236.1 | XP_016856725.1 | 7 | 911 | |||
XM_017001237.1 | XP_016856726.1 | 8 | 1155 | |||
XM_017001238.1 | XP_016856727.1 | 8 | 1094 | |||
XM_017001239.1 | XP_016856728.1 | 8 | 905 | |||
XM_017001240.1 | XP_016856729.1 | 5 | 473 | |||
XM_017001241.1 | XP_016856730.1 | 5 | 621 | |||
XM_017001242.1 | XP_016856731.1 | 5 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
