siRNA Details
Gene Symbol: HLA-J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024240.1 | 4 | 717 | |||
GenBank mRNA | BC020891.1 | - | 3 | 401 | ||
BC062324.1 | - | 2 | 123 | |||
BC089454.1 | - | 3 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
