siRNA Details
Gene Symbol: COL14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021110.3 | NP_066933.1 | 11 | 1507 | ||
XM_005251059.3 | XP_005251116.1 | 11 | 1507 | |||
XM_006716651.3 | XP_006716714.1 | 11 | 1370 | |||
XM_017013809.1 | XP_016869298.1 | 11 | 1296 | |||
GenBank mRNA | AK293080.1 | - | 11 | 1477 | ||
AK307716.1 | - | 11 | 1471 | |||
AK311498.1 | - | 11 | 1471 | |||
BC036192.1 | - | 14 | 1533 | |||
BC140893.1 | - | 10 | 1276 | 9021761 | ||
M64108.1 | - | 6 | 676 | |||
M64109.1 | - | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
