siRNA Details
Gene Symbol: LIN37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019104.2 | NP_061977.1 | 9 | 1068 | ||
GenBank mRNA | AF162447.1 | - | 4 | 432 | ||
AK293116.1 | - | 9 | 833 | |||
BC009071.1 | - | 9 | 787 | 3872616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
