siRNA Details
Gene Symbol: TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145549.2 | NP_001139021.1 | 10 | 1624 | ||
NM_030810.3 | NP_110437.2 | 10 | 1660 | |||
GenBank mRNA | AK025006.1 | - | 4 | 1400 | ||
AK075291.1 | - | 10 | 1615 | |||
AK098089.1 | - | 13 | 1636 | |||
AL833008.1 | - | 10 | 1504 | |||
AL834423.1 | - | 7 | 1058 | |||
AY326464.1 | - | 10 | 1649 | |||
AY358646.1 | - | 10 | 1654 | |||
BC001199.1 | - | 10 | 1360 | 3356029 |
Gene Symbol: BLOC1S5-TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037616.1 | 13 | 1781 | |||
GenBank mRNA | BC052310.1 | - | 13 | 1731 | 6671945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
