siRNA Details
Gene Symbol: TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145549.2 | NP_001139021.1 | 8 | 1002 | ||
NM_030810.3 | NP_110437.2 | 8 | 1038 | |||
GenBank mRNA | AJ440721.1 | - | 8 | 793 | ||
AK025006.1 | - | 2 | 778 | |||
AK075291.1 | - | 8 | 1012 | |||
AK098089.1 | - | 11 | 1014 | |||
AK315598.1 | - | 8 | 1003 | |||
AL833008.1 | - | 8 | 882 | |||
AL834423.1 | - | 5 | 436 | |||
AY326464.1 | - | 8 | 1026 | |||
AY358646.1 | - | 8 | 1032 | |||
BC001199.1 | - | 8 | 757 | 3356029 | ||
BU855393.1 | - | 8 | 821 |
Gene Symbol: BLOC1S5-TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037616.1 | 11 | 1159 | |||
GenBank mRNA | BC052310.1 | - | 11 | 1109 | 6671945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
