siRNA Details
Gene Symbol: TYW1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145440.2 | NP_001138912.2 | 1 | 103 | ||
XM_011516230.2 | XP_011514532.1 | 1 | 103 | |||
XM_011516231.2 | XP_011514533.1 | 1 | 103 | |||
XM_011516232.2 | XP_011514534.1 | 1 | 103 | |||
XM_017012221.1 | XP_016867710.1 | 1 | 103 | |||
XM_017012222.1 | XP_016867711.1 | 1 | 103 | |||
GenBank mRNA | AK301471.1 | - | 1 | 55 | ||
AK303225.1 | - | 1 | 43 | |||
AK309556.1 | - | 1 | 43 | |||
AK310913.1 | - | 1 | 69 | |||
BC068520.1 | - | 1 | 67 | 5276588 | ||
DA769764.1 | - | 1 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
