siRNA Details
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 32 | 4702 | ||
XM_005260768.2 | XP_005260825.1 | 32 | 4717 | |||
XM_006723598.1 | XP_006723661.1 | 33 | 4858 | |||
XM_006723599.1 | XP_006723662.1 | 33 | 4858 | |||
XM_011529309.1 | XP_011527611.1 | 33 | 4858 | |||
XM_011529310.2 | XP_011527612.1 | 31 | 4479 | |||
XM_011529311.2 | XP_011527613.2 | 19 | 4818 | |||
XM_017027972.1 | XP_016883461.1 | 33 | 4858 | |||
XM_017027973.1 | XP_016883462.1 | 30 | 4321 | |||
XM_017027974.1 | XP_016883463.1 | 30 | 4316 | |||
XM_017027975.1 | XP_016883464.1 | 32 | 4650 | |||
XM_017027976.1 | XP_016883465.1 | 30 | 4503 | |||
GenBank mRNA | AK002211.1 | - | 24 | 3571 | ||
AK127228.1 | - | 5 | 2873 | |||
BC171842.1 | - | 30 | 4503 | |||
DQ310704.1 | - | 32 | 4559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
