siRNA Details
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 27 | 3708 | ||
XM_005260768.2 | XP_005260825.1 | 27 | 3723 | |||
XM_006723598.1 | XP_006723661.1 | 28 | 3864 | |||
XM_006723599.1 | XP_006723662.1 | 28 | 3864 | |||
XM_011529309.1 | XP_011527611.1 | 28 | 3864 | |||
XM_011529310.2 | XP_011527612.1 | 26 | 3485 | |||
XM_011529311.2 | XP_011527613.2 | 14 | 3824 | |||
XM_017027972.1 | XP_016883461.1 | 28 | 3864 | |||
XM_017027973.1 | XP_016883462.1 | 25 | 3327 | |||
XM_017027974.1 | XP_016883463.1 | 25 | 3322 | |||
XM_017027975.1 | XP_016883464.1 | 27 | 3656 | |||
XM_017027976.1 | XP_016883465.1 | 25 | 3509 | |||
GenBank mRNA | AK002211.1 | - | 19 | 2577 | ||
BC171842.1 | - | 25 | 3509 | |||
DQ310704.1 | - | 27 | 3565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
