siRNA Details
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 34 | 5125 | ||
XM_005260768.2 | XP_005260825.1 | 34 | 5140 | |||
XM_006723598.1 | XP_006723661.1 | 35 | 5281 | |||
XM_006723599.1 | XP_006723662.1 | 35 | 5281 | |||
XM_011529309.1 | XP_011527611.1 | 35 | 5281 | |||
XM_011529310.2 | XP_011527612.1 | 33 | 4902 | |||
XM_011529311.2 | XP_011527613.2 | 21 | 5241 | |||
XM_017027972.1 | XP_016883461.1 | 35 | 5281 | |||
XM_017027973.1 | XP_016883462.1 | 32 | 4744 | |||
XM_017027974.1 | XP_016883463.1 | 32 | 4739 | |||
XM_017027975.1 | XP_016883464.1 | 34 | 5073 | |||
XM_017027976.1 | XP_016883465.1 | 32 | 4926 | |||
GenBank mRNA | AK002211.1 | - | 26 | 3994 | ||
AK022881.1 | - | 3 | 214 | |||
AK127228.1 | - | 7 | 3296 | |||
BC013749.1 | - | 3 | 273 | |||
BC171842.1 | - | 32 | 4926 | |||
DQ310704.1 | - | 34 | 4982 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
