siRNA Details
Gene Symbol: KRTAP1-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030966.1 | NP_112228.1 | 1 | 926 | ||
GenBank mRNA | AJ406927.1 | - | 1 | 926 | ||
BC069161.1 | - | 1 | 925 | 7216931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
