siRNA Details
Gene Symbol: KRTAP1-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030966.1 | NP_112228.1 | 1 | 588 | ||
GenBank mRNA | AJ406927.1 | - | 1 | 588 | ||
BC069161.1 | - | 1 | 587 | 7216931 | ||
BC101150.1 | - | 1 | 582 | 40015405 | ||
BC101151.2 | - | 1 | 582 | 40015406 | ||
BC101152.1 | - | 1 | 612 | 40015409 | ||
BC101153.2 | - | 1 | 582 | 40015410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
