siRNA Details
Gene Symbol: SLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032444.2 | NP_115820.2 | 12 | 4062 | ||
XM_011522715.2 | XP_011521017.1 | 12 | 4076 | |||
XM_017023775.1 | XP_016879264.1 | 9 | 3467 | |||
GenBank mRNA | AB058687.1 | - | 1436 | |||
BC036335.1 | - | 3 | 1454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
