siRNA Details
Gene Symbol: FBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319193.1 | NP_001306122.1 | 13 | 1234 | ||
GenBank mRNA | AB354594.1 | - | 12 | 1151 | ||
AK074045.1 | - | 13 | 1229 | |||
BC023549.2 | - | 13 | 1370 | 4109753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
