siRNA Details
Gene Symbol: AADACL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013630.1 | NP_001013652.1 | 2 | 266 | ||
XM_017001152.1 | XP_016856641.1 | 2 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
