siRNA Details
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182503.2 | NP_872309.2 | 1 | 88 | ||
XM_011535439.2 | XP_011533741.1 | 1 | 127 | |||
XM_011535440.1 | XP_011533742.1 | 1 | 88 | |||
XM_011535443.1 | XP_011533745.1 | 1 | 134 | |||
XM_011535444.2 | XP_011533746.1 | 1 | 54 | |||
GenBank mRNA | BC037955.1 | - | 1 | 48 | ||
DA100320.1 | - | 1 | 88 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
