siRNA Details
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286259.1 | NP_001273188.1 | 6 | 1896 | ||
NM_182503.2 | NP_872309.2 | 6 | 798 | |||
XM_011535439.2 | XP_011533741.1 | 9 | 954 | |||
XM_011535440.1 | XP_011533742.1 | 9 | 3686 | |||
XM_011535444.2 | XP_011533746.1 | 8 | 817 | |||
XM_017010260.1 | XP_016865749.1 | 7 | 4215 | |||
XM_017010261.1 | XP_016865750.1 | 6 | 4172 | |||
XM_017010262.1 | XP_016865751.1 | 6 | 676 | |||
XM_017010263.1 | XP_016865752.1 | 5 | 650 | |||
GenBank mRNA | AK126201.1 | - | 6 | 1902 | ||
BC037955.1 | - | 6 | 758 | |||
BX538182.1 | - | 6 | 1196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
