siRNA Details
Gene Symbol: RNFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016125.3 | NP_057209.3 | 2 | 528 | ||
GenBank mRNA | AF100745.1 | - | 2 | 536 | ||
AK294806.1 | - | 2 | 509 | |||
AK295162.1 | - | 2 | 484 | |||
AK296197.1 | - | 2 | 509 | |||
BC006971.1 | - | 2 | 506 | |||
BC088365.1 | - | 2 | 491 | 30390062 |
Gene Symbol: TBC1D3P1-DHX40P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002924.3 | 13 | 1585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
