siRNA Details
Gene Symbol: ZNF600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321866.1 | NP_001308795.1 | 1 | 92 | ||
NM_001321867.1 | NP_001308796.1 | 1 | 92 | |||
NM_198457.3 | NP_940859.3 | 1 | 92 | |||
XM_017026390.1 | XP_016881879.1 | 1 | 126 | |||
XM_017026391.1 | XP_016881880.1 | 1 | 126 | |||
GenBank mRNA | AK131231.1 | - | 1 | 82 | ||
BX640933.1 | - | 1 | 64 | |||
DB158392.1 | - | 1 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
