siRNA Details
Gene Symbol: FAM53B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014661.3 | NP_055476.3 | 2 | 484 | ||
GenBank mRNA | AK127343.1 | - | 2 | 678 | ||
BC031654.1 | - | 2 | 392 | 5174078 | ||
BX344666.2 | - | 2 | 482 | |||
CA397342.1 | - | 2 | 397 | |||
D50930.1 | - | 2 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
