siRNA Details
Gene Symbol: FAM53B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014661.3 | NP_055476.3 | 2, 3 | 612 | ||
GenBank mRNA | AK127343.1 | - | 2, 3 | 806 | ||
BC031654.1 | - | 2, 3 | 520 | 5174078 | ||
BX344666.2 | - | 2, 3 | 610 | |||
CA397342.1 | - | 2, 3 | 525 | |||
D50930.1 | - | 2, 3 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
