siRNA Details
Gene Symbol: PSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031246.3 | NP_112536.2 | 5 | 1153 | ||
GenBank mRNA | BC022316.1 | - | 5 | 1080 | 4731250 | |
M20882.1 | - | 5 | 1064 | |||
M37398.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
