siRNA Details
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076678.2 | NP_001070146.1 | 4 | 3695 | ||
NM_175910.6 | NP_787106.4 | 2 | 3472 | |||
GenBank mRNA | AB209086.1 | - | 2 | 3440 | ||
AK095616.1 | - | 1 | 2220 | |||
AK127476.1 | - | 2 | 2499 | |||
AK128494.1 | - | 2 | 3458 | |||
BX648842.1 | - | 4 | 3685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
