siRNA Details
Gene Symbol: CCDC183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039374.4 | NP_001034463.4 | 11 | 1200 | ||
GenBank mRNA | AK090683.1 | - | 6 | 1040 | ||
AK092639.1 | - | 7 | 1723 | |||
AK094220.1 | - | 1 | 886 | |||
BC064582.1 | - | 11 | 1186 | |||
BC137317.1 | - | 11 | 1175 | 9021314 | ||
BC137318.1 | - | 11 | 1175 | 9021315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
