siRNA Details
Gene Symbol: CCDC183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090683.1 | - | 6 | 840 | ||
AK092639.1 | - | 7 | 1522 | |||
AK094220.1 | - | 1 | 685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
