siRNA Details
Gene Symbol: ERI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142725.1 | NP_001136197.1 | 3 | 156 | ||
NM_080663.2 | NP_542394.2 | 3 | 156 | |||
GenBank mRNA | AK127391.1 | - | 3 | 159 | ||
AK292594.1 | - | 4 | 369 | |||
AK304347.1 | - | 3 | 155 | |||
BC010503.1 | - | 3 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
